2-Aminoacetophenon Hydrochlorid, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Phenacylamine hydrochloride

M-Clarity™ Qualitätsstufe = MQ200

A38207-1G A38207-5G
SIALA38207-1GEA 48.1 EUR
SIALA38207-1G SIALA38207-5G
2-Aminoacetophenon Hydrochlorid, Sigma-Aldrich®
2-Aminoacetophenon Hydrochlorid
Formel: C₈H₁₀ClNO
Molecular Weight: 171,63 g/mol
MDL: MFCD00012873
CAS-Nummer: 5468-37-1
EINECS: 226-787-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR