4'-(Methylthio)acetophenon

Lieferant: FLUOROCHEM

Synonyme: 1-(4-(Methylthio)phenyl)ethanone

023852-10G 023852-50G
FLUO023852-10GEA 60.9 EUR
FLUO023852-10G FLUO023852-50G
4'-(Methylthio)acetophenon
4'-(Methylthio)acetophenon
Formel: C₉H₁₀OS
MDL: MFCD00039835
CAS-Nummer: 1778-09-2
EINECS: 217-213-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR