4'-(Trifluormethylthio)acetophenon

Lieferant: CHEMPUR

FL7991.1 FL7991.5
CHMPFL7991.1EA 0 EUR
CHMPFL7991.1 CHMPFL7991.5
4'-(Trifluormethylthio)acetophenon
4'-(Trifluormethylthio)acetophenon
Formel: C₉H₇F₃OS
MDL: MFCD00236332
CAS-Nummer: 713-67-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR