3-Amino-4-(methylthio)benzotrifluorid
Lieferant: FLUOROCHEM
Synonyme:
2-Methylthio-5-(trifluoromethyl)aniline
010401-1G
010401-5G
FLUO010401-1GEA
104
EUR
FLUO010401-1G
FLUO010401-5G
3-Amino-4-(methylthio)benzotrifluorid
3-Amino-4-(methylthio)benzotrifluorid
Formel:
C₈H₈F₃NS Molecular Weight: 207.22 g/mol |
MDL:
MFCD00052715 CAS-Nummer: 207974-07-0 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...