Methyl-5-brom-2-benzo[b]thiophencarboxylat

Lieferant: BIONET RESEARCH

Synonyme: Methyl 5-bromobenzo[b]thiophene-2-carboxylate, Methyl 5-bromo-1-benzothiophene-2-carboxylate

7X-0709.500 7X-0709.1 7X-0709.5 7X-0709.10
BIOT7X-0709.500EA 0 EUR
BIOT7X-0709.500 BIOT7X-0709.1 BIOT7X-0709.5 BIOT7X-0709.10
Methyl-5-brom-2-benzo[b]thiophencarboxylat
Methyl-5-brom-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₇BrO₂S
Molecular Weight: 271.1300 g/mol
MDL: MFCD05662034
CAS-Nummer: 7312-11-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR