Methyl-3-amino-5-nitro-2-benzo[b]thiophencarboxylat

Lieferant: MENAI

CB548.2 CB548.5 CB548.10 CB548.1
MENACB548.2EA 0 EUR
MENACB548.2 MENACB548.5 MENACB548.10 MENACB548.1
Methyl-3-amino-5-nitro-2-benzo[b]thiophencarboxylat
Methyl-3-amino-5-nitro-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₈N₂O₄S
MDL: MFCD00245562
CAS-Nummer: 34674-75-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR