Methyl-3-hydroxy-2-benzo[b]thiophencarboxylat

Lieferant: Thermo Scientific

BTB04875DA BTB04875EA
MAYBBTB04875DAEA 0 EUR
MAYBBTB04875DA MAYBBTB04875EA
Methyl-3-hydroxy-2-benzo[b]thiophencarboxylat
Methyl-3-hydroxy-2-benzo[b]thiophencarboxylat
Formel: C₁₀H₈O₃S
MDL: MFCD00067732
CAS-Nummer: 13134-76-4
EINECS: 236-066-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR