Methyl-3-amino-4-fluor-2-benzo[b]thiophencarboxylat
Lieferant: FLUOROCHEM
Synonyme:
Methyl 3-amino-4-fluorobenzo[b]thiophene-2-carboxylate
021897-5G
021897-25G
FLUO021897-5GEA
125
EUR
FLUO021897-5G
FLUO021897-25G
Methyl-3-amino-4-fluor-2-benzo[b]thiophencarboxylat
Methyl-3-amino-4-fluor-2-benzo[b]thiophencarboxylat
Formel:
C₁₀H₈FNO₂S |
MDL:
MFCD02091511 CAS-Nummer: 144899-95-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...