Hydroxylapatit Type I, Suspension, gepuffert, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Calcium Phosphate, Tribasic
M-Clarity™ Qualitätsstufe = MQ200
H0252-25G
H0252-65G
SIALH0252-25GEA
182
EUR
SIALH0252-25G
SIALH0252-65G
Hydroxylapatit Type I, Suspension, gepuffert, Sigma-Aldrich®
Hydroxylapatit
Formel:
Ca₅HO₁₃P₃ Molecular Weight: 502,32 g/mol |
MDL:
MFCD00010904 CAS-Nummer: 1306-06-5 EINECS: 215-145-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...