Ethylcyclopropancarboxylat ≥98%
Lieferant: Thermo Scientific
|
Gefahr
|
B24870.14
B24870.22
B24870.14EA
44.2
EUR
B24870.14
B24870.22
Ethylcyclopropancarboxylat ≥98%
Ethylcyclopropancarboxylat
Purity: 98%
CAS No.: 4606-07-9
Molecular Formula: C6H10O2
Formula Weight: 114.15
Flash Point: 18°(64°F)
Refractive Index: 1.4200
Boiling Point: 133-134°
Beilstein Reference: 2039752
UN No.: 3272
MDL No.: MFCD00001282
Formel:
C₆H₁₀O₂ Molecular Weight: 114,15 g/mol Siedepunkt: 133…134 °C Dichte: 0,960 Flammpunkt: 18 °C (64 °F) Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001282 CAS-Nummer: 4606-07-9 EINECS: 225-010-4 UN: 3272 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...