Methylcyclopropancarboxylat ≥98%

Lieferant: Thermo Fisher Scientific
Gefahr

Synonyme: Methylcyclopropanecarboxylate

B20853.14 B20853.22 B20853.36
B20853.14EA 39.2 EUR
B20853.14 B20853.22 B20853.36
Methylcyclopropancarboxylat ≥98%
Methylcyclopropancarboxylat

Purity: 98% CAS No.: 2868-37-3 Molecular Formula: C5H8O2 Formula Weight: 100.12 Flash Point: 22°(71°F) Refractive Index: 1.4180 Boiling Point: 118-119° Beilstein Reference: 2039388 UN No.: 3272 MDL No.: MFCD00001280

Formel: C₅H₈O₂
Molecular Weight: 100.12 g/mol
Siedepunkt: 118…119 °C
Dichte: 0.983
Flammpunkt: 22 °C (71 °F)
Lagertemperatur: Raumtemperatur
MDL: MFCD00001280
CAS-Nummer: 2868-37-3
EINECS: 220-690-9
UN: 3272
ADR: 3 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR