Methylcyclopropancarboxylat
Lieferant: FLUOROCHEM
Synonyme:
Methylcyclopropanecarboxylate
022090-100G
022090-1KG
FLUO022090-100GEA
112
EUR
FLUO022090-100G
FLUO022090-1KG
Methylcyclopropancarboxylat
Methylcyclopropancarboxylat
Formel:
C₅H₈O₂ Molecular Weight: 100.12 g/mol |
MDL:
MFCD00001280 CAS-Nummer: 2868-37-3 EINECS: 220-690-9 UN: 3272 ADR: 3 II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...