1-(Trifluormethyl)cyclobutancarbonsäure, Sigma-Aldrich®
Lieferant: Merck
M-Clarity™ Qualitätsstufe = MQ100
665592-100MG
665592-500MG
SIAL665592-100MGEA
85.7
EUR
SIAL665592-100MG
SIAL665592-500MG
1-(Trifluormethyl)cyclobutancarbonsäure, Sigma-Aldrich®
1-(Trifluoromethyl)cyclobutanecarboxylic acid
Formel:
C₆H₇F₃O₂ Molecular Weight: 168,12 g/mol |
MDL:
MFCD03425197 CAS-Nummer: 277756-45-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...