1-(Trifluormethyl)cyclopentancarbonsäure, Sigma-Aldrich®

Lieferant: Merck


M-Clarity™ Qualitätsstufe = MQ100

694398-1G 694398-250MG
SIAL694398-1GEA 213 EUR
SIAL694398-1G SIAL694398-250MG
1-(Trifluormethyl)cyclopentancarbonsäure, Sigma-Aldrich®
1-(Trifluoromethyl)cyclopentanecarboxylic acid
Formel: C₇H₉F₃O₂
Molecular Weight: 182,15 g/mol
MDL: MFCD08445821
CAS-Nummer: 277756-44-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR