1-Cyclohexen-1-carbonsäure, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cyclohex-1-enecarboxylic acid
M-Clarity™ Qualitätsstufe = MQ100
328367-1G
328367-5G
SIAL328367-1GEA
30.2
EUR
SIAL328367-1G
SIAL328367-5G
1-Cyclohexen-1-carbonsäure, Sigma-Aldrich®
1-Cyclohexen-1-carbonsäure
Formel:
C₇H₁₀O₂ Molecular Weight: 126,16 g/mol |
MDL:
MFCD00001545 CAS-Nummer: 636-82-8 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...