1-Cyclopenten-1-carbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 1-Cyclopentenecarboxylic acid

328324-1G 328324-5G
SIAL328324-1GEA 35.4 EUR
SIAL328324-1G SIAL328324-5G
1-Cyclopenten-1-carbonsäure, Sigma-Aldrich®
1-Cyclopenten-1-carbonsäure
Formel: C₆H₈O₂
Molecular Weight: 112,13 g/mol
MDL: MFCD00001396
CAS-Nummer: 1560-11-8
UN: 3261
ADR: 8,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR