5-Fluor-3-methylbenzo[b]thiophen

Lieferant: FLUOROCHEM

008343-1G 008343-5G
FLUO008343-1GEA 71.9 EUR
FLUO008343-1G FLUO008343-5G
5-Fluor-3-methylbenzo[b]thiophen
5-Fluor-3-methylbenzo[b]thiophen
Formel: C₉H₇FS
MDL: MFCD00179348
CAS-Nummer: 17514-63-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR