1-Phenylcyclopropancarbonsäure 97%

Lieferant: Thermo Scientific

Synonyme: 1-Phenyl-1-cyclopropanecarboxylic acid

170680100 170680500
ACRO170680100EA 83.7 EUR
ACRO170680100 ACRO170680500
1-Phenylcyclopropancarbonsäure 97%
1-Phenylcyclopropanecarboxylic acid
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
Schmelzpunkt: 84…87 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00001288
CAS-Nummer: 6120-95-2
EINECS: 228-090-9

Bestellen

Spezifikation Testergebnisse

Appearance White to beige Crystalline powder or crystals and/or chunks
Infrared spectrum Conforms
Melting point 84 to 87 °C
GC ≥96.0 %
Water ≤0.5 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR