1-Phenylcyclopropancarbonsäure ≥97%
Lieferant: Thermo Fisher Scientific
|
Achtung
|
Synonyme:
1-Phenyl-1-cyclopropanecarboxylic acid
H27525.14
H27525.06
H27525.14EA
173
EUR
H27525.14
H27525.06
1-Phenylcyclopropancarbonsäure ≥97%
1-Phenylcyclopropanecarboxylic acid
Purity: 97%
CAS No.: 6120-95-2
Molecular Formula: C10H10O2
Formula Weight: 162.19
Melting Point: 85-89°
MDL No.: MFCD00001288
Formel:
C₁₀H₁₀O₂ Molecular Weight: 162.19 g/mol Schmelzpunkt: 85…89 °C Lagertemperatur: Raumtemperatur |
MDL:
MFCD00001288 CAS-Nummer: 6120-95-2 EINECS: 228-090-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...