1-Phenylcyclopropancarbonsäure

Lieferant: COMBI-BLOCKS

Synonyme: 1-Phenyl-1-cyclopropanecarboxylic acid

HC-2889-25G HC-2889-1G HC-2889-5G
COBBHC-2889-25GEA 182 EUR
COBBHC-2889-25G COBBHC-2889-1G COBBHC-2889-5G
1-Phenylcyclopropancarbonsäure
1-Phenylcyclopropanecarboxylic acid
Formel: C₁₀H₁₀O₂
Molecular Weight: 162,19 g/mol
MDL: MFCD00001288
CAS-Nummer: 6120-95-2
EINECS: 228-090-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR