trans-2-Phenylcyclopropancarbonsäure, Sigma-Aldrich®

Lieferant: Merck

Synonyme: trans-2-Phenylcyclopropane-1-carboxylic acid

M-Clarity™ Qualitätsstufe = MQ100

P22354-25G P22354-5G
SIALP22354-25GEA 251 EUR
SIALP22354-25G SIALP22354-5G
trans-2-Phenylcyclopropancarbonsäure, Sigma-Aldrich®
trans-2-Phenylcyclopropanecarboxylic acid
Formel: C₁₀H₁₀O₂
Molecular Weight: 162.1852 g/mol
MDL: MFCD00001292
CAS-Nummer: 939-90-2
EINECS: 213-366-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR