Benzo[a]pyren-[D12] (98% D)

Lieferant: LGC Standards PROMOCHEM
Gefahr

Synonyme: Benzo[a]pyrene-[²H₁₂]

CILDLM2580.01 CILDLM2580.05
PROOCILDLM2580.01EA 310 EUR
PROOCILDLM2580.01 PROOCILDLM2580.05
Benzo[a]pyren-[D12] (98% D)
Benzo[a]pyren-[D12]
Formel: C₂₀D₁₂
CAS-Nummer: 63466-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR