N-Fmoc-D-leucin ≥98%
Lieferant: Thermo Scientific
Synonyme:
Fmoc-D-Leu-OH
H29041.06
H29041.14
H29041.03
H29041.06EA
81.6
EUR
H29041.06
H29041.14
H29041.03
N-Fmoc-D-leucin ≥98%
N-Fmoc-D-leucin
Purity: 98%
CAS No.: 114360-54-2
Molecular Formula: C21H23NO4
Formula Weight: 353.42
Storage Temperature: 2°C to 8°C
Melting Point: 151-154°
Optical Rotation: +25° (c=1 in DMF)
MDL No.: MFCD00062957
Formel:
C₂₁H₂₃NO₄ Molecular Weight: 353,4 g/mol Schmelzpunkt: 151…154 °C |
MDL:
MFCD00062957 CAS-Nummer: 114360-54-2 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...