N-Fmoc-D-leucin

Lieferant: NOVABIOCHEM

Synonyme: Fmoc-D-Leu-OH

8.52145.0025 8.52145.0005
04-13-1025-25EA 393 EUR
04-13-1025-25 04-13-1025-5
N-Fmoc-D-leucin
N-Fmoc-D-leucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,4 g/mol
MDL: MFCD00062957
CAS-Nummer: 114360-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR