N-Fmoc-D-leucin, Sigma-Aldrich®

Lieferant: Merck

Synonyme: Fmoc-D-Leu-OH

47316-1G 47316-5G
SIAL47316-1GEA 26 EUR
SIAL47316-1G SIAL47316-5G
N-Fmoc-D-leucin, Sigma-Aldrich®
N-Fmoc-D-leucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,4 g/mol
MDL: MFCD00062957
CAS-Nummer: 114360-54-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR