N-Fmoc-L-leucin

Lieferant: NOVABIOCHEM

8.52011.0100 8.52011.0025
04-12-1025-100EA 67.9 EUR
04-12-1025-100 04-12-1025-25
N-Fmoc-L-leucin
N-Fmoc-L-leucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353,4 g/mol
MDL: MFCD00037133
CAS-Nummer: 35661-60-0
EINECS: 252-662-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR