N-Fmoc-L-leucin, Sigma-Aldrich®

Lieferant: Merck

47633-50G-F 47633-5G-F
SIAL47633-50G-FEA 76.8 EUR
SIAL47633-50G-F SIAL47633-5G-F
N-Fmoc-L-leucin, Sigma-Aldrich®
N-Fmoc-L-leucin
Formel: C₂₁H₂₃NO₄
Molecular Weight: 353.4 g/mol
MDL: MFCD00037133
CAS-Nummer: 35661-60-0
EINECS: 252-662-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR