Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid) ≥98%
Lieferant: Thermo Scientific
Achtung
Synonyme: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide
A13143.03 A13143.06 A13143.14
A13143.03EA 21.1 EUR
A13143.03 A13143.06 A13143.14
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid) ≥98%
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Purity: 98% CAS No.: 82911-69-1 Molecular Formula: C19H15NO5 Formula Weight: 337.34 Storage Temperature: 2°C to 8°C Melting Point: 150-153° Beilstein Reference: 3569540 UN No.: 3077 MDL No.: MFCD00010733
Formel: C₁₉H₁₅NO₅
Molecular Weight: 337,3 g/mol
Schmelzpunkt: 150…153 °C
MDL: MFCD00010733
CAS-Nummer: 82911-69-1
EINECS: 433-520-5
ADR: 9,III