Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)

Lieferant: BACHEM BIOCHEMICA

Synonyme: Fmoc-OSu, N-(9-Fluorenylmethoxycarbonyloxy)succinimide

4006805.0025 4006805.0100 4006805.0250
BACE4006805.0025EA 161 EUR
BACE4006805.0025 BACE4006805.0100 BACE4006805.0250
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Fmoc-OSu (N-(9-Fluorenylmethoxycarbonyloxy)succinimid)
Formel: C₁₉H₁₅NO₅
Molecular Weight: 337.3 g/mol
MDL: MFCD00010733
CAS-Nummer: 82911-69-1
EINECS: 433-520-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR