2,4-Dimethyl-3-ethylpyrrol, Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Cryptopyrrole, 3-Ethyl-2,4-dimethylpyrrole, Kryptopyrrole
M-Clarity™ Qualitätsstufe = MQ100
D158402-25G
D158402-5G
SIALD158402-25GEA
370
EUR
SIALD158402-25G
SIALD158402-5G
2,4-Dimethyl-3-ethylpyrrol, Sigma-Aldrich®
2,4-Dimethyl-3-ethylpyrrole
Formel:
C₈H₁₃N Molecular Weight: 123.2 g/mol |
MDL:
MFCD00005222 CAS-Nummer: 517-22-6 EINECS: 208-234-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...