Storage tubes, MATRIX™
Lieferant: Thermo Fisher Scientific
Matrix™
MATR4271EA 263 EUR
MATR4271 MATR4150 MATR4272 MATR4140 MATR4250 MATR4170 MATR4253 MATR4273 MATR4251 MATR4252 MATR4274 MATR4147 MATR4148 MATR4247 MATR4248 MATR4249
Storage tubes, MATRIX™
Röhrchen Reagenzgläser- und Probenröhrchen
PP, translucent
  • Non sterile or sterile available
  • Tubes are 100% leak tested to ensure sample integrity and security
  • Tubes are validated to be free of DNAse, RNAse, endotoxins and cytotoxins
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or deep well block sample storage.
Store valuable samples where 2D tracking is not required with high-density Thermo Scientific™ Matrix Blank and Alphanumeric Storage Tubes, offered in manual and automation-friendly racks. Visual identification of samples can be made using the alphanumeric code on the bottom of each tube, and the tubes are compatible with the full line of seals and capping equipment. Ideal for scaling up from microplate or deep well block sample storage.