2-Chlor-4-pyridinboronsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Chlorpyridin-4-boronsäure, 2-Chlor-4-pyridinylboronsäure

016271-1G 016271-5G
FLUO016271-1GEA 76.2 EUR
FLUO016271-1G FLUO016271-5G
2-Chlor-4-pyridinboronsäure
2-Chloro-4-pyridineboronic acid
Formel: C₅H₅BClNO₂
Molecular Weight: 157.36 g/mol
MDL: MFCD03788416
CAS-Nummer: 458532-96-2

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR