2,4-Dichlor-N-methylanilin

Lieferant: COMBI-BLOCKS

HC-2855-5G HC-2855-25G HC-2855-1G
COBBHC-2855-5GEA 149 EUR
COBBHC-2855-5G COBBHC-2855-25G COBBHC-2855-1G
2,4-Dichlor-N-methylanilin
2,4-Dichlor-N-methylanilin
Formel: C₇H₇Cl₂N
Molecular Weight: 176,04 g/mol
MDL: MFCD00052853
CAS-Nummer: 35113-88-3
UN: 2811
ADR: 6.1,III

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR