6-Fluornicotinsäure

Lieferant: COMBI-BLOCKS

Synonyme: 2-Fluoro-5-pyridinecarboxylic acid

PY-7066-1G PY-7066-25G PY-7066-5G
COBBPY-7066-1GEA 25.5 EUR
COBBPY-7066-1G COBBPY-7066-25G COBBPY-7066-5G
6-Fluornicotinsäure
6-Fluornicotinsäure
Formel: C₆H₄FNO₂
Molecular Weight: 141,1 g/mol
MDL: MFCD01859863
CAS-Nummer: 403-45-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR