2-(Propylthio)nicotinsäure
Lieferant: Thermo Scientific
Synonyme:
2-(n-Propylthio)nicotinic acid
BTB06368DA
BTB06368EA
BTB06368.50
MAYBBTB06368DAEA
0
EUR
MAYBBTB06368DA
MAYBBTB06368EA
MAYBBTB06368.50
2-(Propylthio)nicotinsäure
2-(Propylthio)nicotinsäure
Formel:
C₉H₁₁NO₂S Molecular Weight: 197,26 g/mol |
MDL:
MFCD00051682 CAS-Nummer: 175135-22-5 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...