2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin
Lieferant: FLUOROCHEM
Synonyme:
4-Aminotetrafluorobenzotrifluoride
008618-5G
008618-1G
FLUO008618-5GEA
267
EUR
FLUO008618-5G
FLUO008618-1G
2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin
2,3,5,6-Tetrafluor-4-(trifluormethyl)anilin
Formel:
C₇H₂F₇N Molecular Weight: 233.09 g/mol |
MDL:
MFCD00091518 CAS-Nummer: 651-83-2 UN: 2810 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...