2-(4-Chlorphenyl)thioacetamid

Lieferant: FLUOROCHEM

021352-1G 021352-5G
FLUO021352-1GEA 0 EUR
FLUO021352-1G FLUO021352-5G
2-(4-Chlorphenyl)thioacetamid
2-(4-Chlorphenyl)thioacetamid
Formel: C₈H₈ClNS
Molecular Weight: 185,67 g/mol
MDL: MFCD00278249
CAS-Nummer: 17518-48-8

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR