3-Amino-4-(4-morpholino)benzotrifluorid

Lieferant: MENAI

Synonyme: 2-(4-Morpholinyl)-5-(trifluoromethyl)aniline, 2-Morpholino-5-(trifluoromethyl)aniline

WB141.5 WB141.10
MENAWB141.5EA 0 EUR
MENAWB141.5 MENAWB141.10
3-Amino-4-(4-morpholino)benzotrifluorid
3-Amino-4-(4-morpholino)benzotrifluorid
Formel: C₁₁H₁₃F₃N₂O
Molecular Weight: 246.23 g/mol
MDL: MFCD00042160
CAS-Nummer: 784-57-6
UN: 2811
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR