N-(Cyanacetyl)morpholin

Lieferant: FLUOROCHEM

Synonyme: 3-Morpholino-3-oxopropanenitrile, 3-(Morpholin-4-yl)-3-oxopropanenitrile

CFF187.25 CFF187.50 017256-25G 017256-50G
CHMPCFF187.25EA 268 EUR
CHMPCFF187.25 CHMPCFF187.50 FLUO017256-25G FLUO017256-50G
N-(Cyanacetyl)morpholin
N-(Cyanacetyl)morpholin
Formel: C₇H₁₀N₂O₂
Molecular Weight: 154,17 g/mol
MDL: MFCD00023353
CAS-Nummer: 15029-32-0
EINECS: 239-112-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR