6-Brom-3-formylchromon 97%

Lieferant: Thermo Scientific
Achtung

Synonyme: 6-Bromo-4-oxo-4H-chromene-3-carbaldehyde

H32053.03 H32053.06
H32053.03EA 17.5 EUR
H32053.03 H32053.06
6-Brom-3-formylchromon 97%
6-Brom-3-formylchromon

Purity: 97% CAS No.: 52817-12-6 Molecular Formula: C10H5BrO3 Formula Weight: 253.05 Melting Point: 190-193° MDL No.: MFCD00191849

Formel: C₁₀H₅BrO₃
Molecular Weight: 253,05 g/mol
Schmelzpunkt: 190…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD00191849
CAS-Nummer: 52817-12-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR