6-Brom-3-cyanchromon

Lieferant: CHEMPUR

Synonyme: 6-Bromo-3-cyanochromone, 6-Bromo-4-oxo-4H-1-benzopyran-3-carbonitrile, 6-Bromochromone-3-carbonitrile, 6-Bromo-4-oxo-4H-chromene-3-carbonitrile

FL12018.POR 012018-5G 012018-1G
CHMPFL12018.POREA 0 EUR
CHMPFL12018.POR FLUO012018-5G FLUO012018-1G
6-Brom-3-cyanchromon
6-Brom-3-cyanchromon
Formel: C₁₀H₄BrNO₂
Molecular Weight: 250,483 g/mol
MDL: MFCD00191842
CAS-Nummer: 52817-13-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR