6-Bromchromon-3-carbonsäure 97%

Lieferant: Thermo Fisher Scientific
Achtung

H31905.03 H31905.06
H31905.03EA 59.5 EUR
H31905.03 H31905.06
6-Bromchromon-3-carbonsäure 97%
6-Bromchromon-3-carbonsäure

Purity: 97% CAS No.: 51085-91-7 Molecular Formula: C10H5BrO4 Formula Weight: 269.05 Melting Point: 191-193° MDL No.: MFCD01548933

Formel: C₁₀H₅BrO₄
Molecular Weight: 269.05 g/mol
Schmelzpunkt: 191…193 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD01548933
CAS-Nummer: 51085-91-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR