6-Bromchromon-3-carbonsäure
Lieferant: CHEMPUR
FL12044.POR
012044-5G
012044-1G
CHMPFL12044.POREA
0
EUR
CHMPFL12044.POR
FLUO012044-5G
FLUO012044-1G
6-Bromchromon-3-carbonsäure
6-Bromchromon-3-carbonsäure
Formel:
C₁₀H₅BrO₄ Molecular Weight: 269.05 g/mol |
MDL:
MFCD01548933 CAS-Nummer: 51085-91-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...