Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on), Sigma-Aldrich®
Lieferant: Merck
Synonyme:
Scopoletin, 7-Hydroxy-6-methoxy-2H-chromen-2-one
M-Clarity™ Qualitätsstufe = MQ100
S2500-100MG
S2500-1G
S2500-500MG
S2500-50MG
SIALS2500-100MGEA
286
EUR
SIALS2500-100MG
SIALS2500-1G
SIALS2500-500MG
SIALS2500-50MG
Scopoletin (7-Hydroxy-6-methoxy-2H-chromen-2-on), Sigma-Aldrich®
Scopoletin (7-Hydroxy-6-methoxycumarin)
Formel:
C₁₀H₈O₄ Molecular Weight: 192,1681 g/mol |
MDL:
MFCD00006872 CAS-Nummer: 92-61-5 EINECS: 202-171-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...