6-Fluorchromon 97%

Lieferant: Thermo Scientific

Synonyme: 6-Fluoro-4H-chromen-4-one

H31907.03 H31907.06
H31907.03EA 33 EUR
H31907.03 H31907.06
6-Fluorchromon 97%
6-Fluorchromon

Purity: 97% CAS No.: 105300-38-7 Molecular Formula: C9H5FO2 Formula Weight: 164.14 Melting Point: 166-169° MDL No.: MFCD03094002

Formel: C₉H₅FO₂
Molecular Weight: 164,14 g/mol
Schmelzpunkt: 166…169 °C
Lagertemperatur: Raumtemperatur
MDL: MFCD03094002
CAS-Nummer: 105300-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR