6-Fluorchromon, Sigma-Aldrich®

Lieferant: Merck

Synonyme: 6-Fluoro-4H-chromen-4-one

M-Clarity™ Qualitätsstufe = MQ100

543659-1G 543659-5G
SIAL543659-1GEA 74 EUR
SIAL543659-1G SIAL543659-5G
6-Fluorchromon, Sigma-Aldrich®
6-Fluorchromon
Formel: C₉H₅FO₂
Molecular Weight: 164,14 g/mol
MDL: MFCD03094002
CAS-Nummer: 105300-38-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR