2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd

Lieferant: BIONET RESEARCH

Synonyme: 2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde

12R-0210.500 12R-0210.1 12R-0210.5
BIOT12R-0210.500EA 0 EUR
BIOT12R-0210.500 BIOT12R-0210.1 BIOT12R-0210.5
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
Formel: C₅H₃Cl₂N₃O
Molecular Weight: 192,00 g/mol
MDL: MFCD03001242
CAS-Nummer: 5604-46-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR