2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
Lieferant: FLUOROCHEM
Synonyme:
2-Amino-4,6-dichloropyrimidine-5-carboxaldehyde
011354-10G
011354-1G
011354-5G
FLUO011354-10GEA
165
EUR
FLUO011354-10G
FLUO011354-1G
FLUO011354-5G
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
2-Amino-4,6-dichlorpyrimidin-5-carbaldehyd
Formel:
C₅H₃Cl₂N₃O Molecular Weight: 192.00 g/mol |
MDL:
MFCD03001242 CAS-Nummer: 5604-46-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...