Cobalt(II)oxalat Dihydrat ≥99,995% (Metall-Basis), Puratronic®

Lieferant: Thermo Fisher Scientific
Achtung

Synonyme: Cobalt(II) oxalate dihydrate

13012.09 13012.18
13012.09EA 116 EUR
13012.09 13012.18
Cobalt(II)oxalat Dihydrat ≥99,995% (Metall-Basis), Puratronic®
Cobalt(II)oxalat Dihydrat

Purity: 99.995% CAS No.: 5965-38-8 Molecular Formula: C2CoO4.2H2O Formula Weight: 182.98 (146.95anhy) Physical Form: Powder Density: 3.021 Appearance: Pink Odour: Odourless Melting Point: 229-231° dec. Merck Reference: 14,2445 UN No.: 3288 MDL No.: MFCD00050793

Formel: C₂CoO₄·2H₂O
Schmelzpunkt: 229…231 °C (Zers.)
Dichte: 3,021
Lagertemperatur: Raumtemperatur
MDL: MFCD03414148
CAS-Nummer: 5965-38-8
EINECS: 212-409-3
UN: 3288
ADR: 6.1,III
Merck Index: 14,2445

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR