5-Methyl-3-isoxazolcarbonylchlorid
Lieferant: FLUOROCHEM
CFF302.5
CFF302.10
CFF302.25
017344-5G
017344-1G
CHMPCFF302.5EA
508
EUR
CHMPCFF302.5
CHMPCFF302.10
CHMPCFF302.25
FLUO017344-5G
FLUO017344-1G
5-Methyl-3-isoxazolcarbonylchlorid
5-Methyl-3-isoxazolcarbonylchlorid
Formel:
C₅H₄ClNO₂ Molecular Weight: 145,55 g/mol |
MDL:
MFCD00085127 CAS-Nummer: 39499-34-8 EINECS: 254-475-6 UN: 3265 ADR: 8,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...